Learn More
Description
The EpiJET 5-hmC and 5-mC Analysis Kit includes three advanced enzymes: T4 phage β-glucosyltransferase (T4 BGT), Epi MspI and Epi HpaII. The Epi MspI and Epi HpaII are isoschisomers with differing sensitivity to internal C methylation of the CCGG site. Epi MspI completely digests genomic DNA when cytosine, 5-methylcytosine (5-mC) or 5-hydroxymethylcytosine (5-hmC) is within its recognition sequence, but its activity is completely blocked when 5-hmC is glucosylated. Epi HpaII cleaves the unmodified CCGG site only.
The kit takes advantage of the T4 BGT ability to specifically modify 5-hmC residues by adding glucose moiety to 5-hmC and the sensitivity of restriction enzyme Epi MspI to glucosylated 5-hmC. The T4 BGT is formulated for highly specific, complete and fast glucosylation of 5-hmC in 15 minutes.
The analysis involves DNA sample glucosylation and subsequent restriction digestion with Epi MspI and Epi HpaII. Restriction digestion products are then analyzed by qPCR with a primer pair flanking CCGG site of interest.
Highlights
- Efficient and reliable - accurate quantification of 5-hmC and 5-mC
- Robust - broad range (from 0.2μg to 4μg) of input DNA
- Fast - procedure completed in 5 hours, no overnight incubation
Applications
- 5-hmC and 5-mC analysis and quantification in CCGG sequences
Includes
- T4 β - glucosyltransferase, 5U/μL
- 10X Epi Buffer
- 10X UDP–glucose
- Epi MspI
- Epi HpaII
- 5-hmC Control DNA
- 5-mC Control DNA
- Unmodified Control DNA
- Control primer 1
- Control primer 2
- Water, nuclease-free
- Detailed Protocol
Specifications
Specifications
| Compatible Buffer | 10X Epi Buffer |
| Quantity | 25 reactions |
| Product Type | 5-hmC and 5-mC Analysis Kit |
| Product Line | EpiJET |
Frequently Asked Questions (FAQs)
The new EpiJET 5-hmC and 5-mC Analysis Kit is more universal than the previous EpiJET 5-hmC Analysis Kit because it provides a total assessment of both cytosine modifications - 5-hydroxymethylcytosine (5-hmC) and methylcytosine (5-mC) - within specific DNA loci (for comparison - the current kit allows only 5-hmC analysis). Additionally, all necessary DNA controls do not originate from plasmids widely used in molecular biology laboratories are included into the new kit.
For each glucosylation reaction, a broad range of DNA input can be used from 50 ng to 1 µg.
DNA purity is an important factor for successful digestion by Epi MspI/HpaII. We recommend use of spin column-based kits such as the Thermo Scientific GeneJET Genomic DNA Purification Kit (Cat. No. K0721/2) for genomic DNA purification.
Yes, Thermo Scientific T4 B-glucosyltransferase is available as a stand-alone enzyme (Cat. No. EO0831, 500 U).
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.