Learn More
Thermo Scientific™ Wash Buffer 1 for MagJET Blood RNA Kit (concentrated)
Description
The Thermo Scientific™ MagJET™ Whole Blood RNA Kit is designed for fast and efficient purification of total RNA from fresh or frozen whole blood treated with EDTA or citrate, blood fractions, bone marrow, or samples stabilized with PAXgene™ or Tempus™. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Specifications
| Buffer | Wash Buffer |
| Product Line | MagJET |
| Quantity | 80 mL |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.