Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffers for MagJET™ Plant gDNA Kit

Catalog No. FERR2481
Change view
Click to view available options
Quantity:
32 mL
45 mL
60 mL
70 mL
225 mL
Product Type:
Elution Buffer
Lysis Buffer A
Lysis Buffer B
Wash Buffer 1
Wash Buffer 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
FERR2481 60 mL Lysis Buffer A
FERR2482 225 mL Lysis Buffer A
FERR2492 32 mL Lysis Buffer B
FERR2511 45 mL Wash Buffer 1
FERR2531 70 mL Elution Buffer
FERR2521 60 mL Wash Buffer 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. FERR2481 Supplier Thermo Scientific™ Supplier No. R2481

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Elute purified plant gDNA for use in common molecular biology procedures. Thermo Scientific™ MagJET™ Elution Buffer is a component of the MagJET™ Plant gDNA Kit (K2761) and may be purchased separately when additional solution is needed. It is provided in 70mL aliquots.

The Thermo Scientific™ MagJET™ Plant Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MagJET Plant gDNA Kit
Product Type Lysis Buffer A
Quantity 60 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.