Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Rebinding Buffer for MagJET Plant RNA Kit

Catalog No. FERR2572
Change view
Click to view available options
Quantity:
20 mL
70 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR2572 70 mL
FERR2571 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR2572 Supplier Thermo Scientific™ Supplier No. R2572

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

The Thermo Scientific™ MagJET™ Plant RNA Kit is designed for fast and efficient purification of total RNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Rebinding Buffer
For Use With (Equipment) MagJET Plant RNA Kit
Product Line MagJET
Quantity 70 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.