Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffers for MagJET™ Viral Kit

Catalog No. FERR2642
Change view
Click to view available options
Quantity:
45 mL
50 mL
155 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR2642 155 mL
FERR2651 45 mL
FERR2661 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FERR2642 Supplier Thermo Scientific™ Supplier No. R2642

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Wash lysed samples to remove any contaminants or inhibitors from the isolated nucleic acids. Thermo Scientific™ MagJET™ Wash Buffer 2 (concentrated) is a component of the MagJET™ Viral DNA and RNA Kits (K2781/2782) and may be purchased separately in 50mL aliquots.

The Thermo Scientific™ MagJET™ Viral DNA and RNA Kit is designed for fast and efficient purification of viral nucleic acids from human and animal samples, including plasma, serum, saliva, and urine. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA and RNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MagJET Viral Kit
Quantity 155 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.