Learn More
Thermo Scientific™ FirstDefender™ RMX Handheld Chemical Identification
Designed for flexible use modes; uses Raman spectroscopy to quickly identify unknown solid and liquid chemicals down range
Supplier: Thermo Scientific™ 8000021201
Description
- Sophisticated chemometric algorithms automatically determine presence of mixed and contaminated chemicals
- Flexible modes—offers handheld use with fixed probe or vial, or easily connects to a tactical robot through integrated RS232 port for universal control
- Designed for use down range with a tough form factor, quick decontamination and easy operation in bulky protective gear
- Lightweight, ergonomically designed unit weighs 2 lb. (919g) for easy transportation down range
- Point-and-shoot sampling offers improved operation through sealed translucent containers; avoids exposure and maintains evidence
- Noncontact analysis requires no consumables and no calibration
- The easy-to-navigate vast substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
- Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
- Can be powered using the included rechargeable battery, wall adapter or SureFire™ disposable batteries
Certifications
MIL-STD 810G and IP67 certified.
Specifications
2-year warranty and support |