Learn More
Thermo Scientific™ Fixed Needle Autosampler Syringes, 1000uL, for Thermo Scientific™ Instruments
Catalog No. 03052758 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
1 mL
Catalog No. Volume (Metric)
03-052-758 1 mL
1 options
Catalog No. 03-052-758 Supplier Thermo Scientific™ Supplier No. 365Q2121
Add to Cart
Add to Cart
Gas-tight syringe, headspace
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 23 G
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Volume (Metric) 1 mL
Length (Metric) Needle 65 mm
Needle Point Style Side Hole
For Use With (Equipment) TriPlus RSH Autosampler