Learn More
Thermo Scientific™ GLP Compliance Services for Plasma Blast Freezer
Description
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific Blast Freezer including all Thermo Scientific XBF |
| Description | Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | Includes IQ/OQ Documentation |