Learn More
Thermo Scientific™ High Purity Single Stage Brass Gas Regulators
Description
Includes
Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
- For non-corrosive analytical and process applications
- Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
- Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
- Capable of withstanding an internal vacuum
- All regulators supplied with 1/8 in. stainless steel output adaptor
- Built in check valve allows for cylinder change without contaminating the gas line
Specifications
Specifications
| Range | 0 to 200 psig |
| Type | CGA 350 with Triple Filter |