Learn More
Thermo Scientific™ High Purity Single Stage Brass Gas Regulators
Catalog No. 60180919
Change view
Click to view available options
Type:
CGA 320 with Combi Filter
CGA 320 with Triple Filter
CGA 350 with Combi Filter
CGA 350 with Triple Filter
CGA 580 with Combi Filter
CGA 580 with Triple Filter
DIN 1 with Combi Filter
DIN 6 with Combi Filter
DIN 6 with Triple Filter
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
60-180-919 CGA 350 with Combi Filter
60-180-901 CGA 580 with Triple Filter
60-180-903 DIN 6 with Triple Filter
60-180-905 CGA 350 with Triple Filter
60-180-909 CGA 320 with Triple Filter
60-180-915 CGA 580 with Combi Filter
60-180-917 DIN 6 with Combi Filter
60-180-921 DIN 1 with Combi Filter
60-180-923 CGA 320 with Combi Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60-180-919 Supplier Thermo Scientific™ Supplier No. 60180919
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Recommended for applications where precise flow control is not critical
  • For non-corrosive analytical and process applications
  • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
  • Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
  • Capable of withstanding an internal vacuum
  • All regulators supplied with 1/8 in. stainless steel output adaptor
  • Built in check valve allows for cylinder change without contaminating the gas line
TRUSTED_SUSTAINABILITY
Specifications
Range 0 to 200 psig
Type CGA 350 with Combi Filter