Learn More
Thermo Scientific™ High Purity Two Stage Brass Gas Regulators
Catalog No. 60180924
Change view
Click to view available options
Type:
CGA 320 with Combi Filter
CGA 320 with Triple Filter
CGA 350 with Combi Filter
CGA 580 with Triple Filter
CGA 590 with Combi Filter
DIN 1 with Combi Filter
DIN 1 with Triple Filter
DIN 6 with Combi Filter
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
60-180-924 CGA 320 with Combi Filter
60-180-902 CGA 580 with Triple Filter
60-180-908 DIN 1 with Triple Filter
60-180-910 CGA 320 with Triple Filter
60-180-918 DIN 6 with Combi Filter
60-180-922 DIN 1 with Combi Filter
60-180-920 CGA 350 with Combi Filter
60-180-926 CGA 590 with Combi Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 60-180-924 Supplier Thermo Scientific™ Supplier No. 60180924
Includes
Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Recommended for non-corrosive analytical and process applications requiring precise, stable, delivery pressure control
  • For non-corrosive analytical and process applications
  • Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
  • Specially designed, convoluted, stainless steel diaphragm provides goodregulating performance and maximum purity integrity without the needfor a soft seal
  • Capable of withstanding an internal vacuum
  • All regulators supplied with 1/8 in. stainless steel output adaptor
  • Built in check valve allows for cylinder change without contaminating the gas line
TRUSTED_SUSTAINABILITY
Specifications
Range 0 to 200 psig
Type CGA 320 with Combi Filter