Learn More
Thermo Scientific™ HPLC Syringes for CTC Instruments
Specifications
Specifications
| Needle Gauge | 22 G |
| Length (English) Needle | 1.9 in. |
| Product Type | HPLC Syringe |
| Quantity | Ea. |
| Volume (Metric) | 10 μL |
| For Use With (Equipment) | CTC Instruments |
| Connection Type | Fixed |