Learn More
Thermo Scientific™ Hybaid EasiSeal Frames
Catalog No. HBOSSSEZ4E Shop All Thermo Scientific Products
Change view
Click to view available options
EasiSeal, Extra Large
Catalog No.
HBOSSSEZ4E EasiSeal, Extra Large
1 options
Catalog No. HBOSSSEZ4E Supplier Thermo Scientific™ Supplier No. HBOSSSEZ4E
Prevent evaporation during incubations on slides
  • Hold reaction mix during hybridization step of in situ PCR
  • Consist of square or rectangular adhesive barrier frame and tabbed cover slip
  • Easily removed from slides after incubation