Learn More
Thermo Scientific™ Hybaid OmniSlide Thermal Cycler System
Catalog No. HBOSBB Shop All Thermo Scientific Products
Change view
Click to view available options
Omnislide Flatblock Unit
Catalog No.
HBOSBB Omnislide Flatblock Unit
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. HBOSBB Supplier Thermo Scientific™ Supplier No. HBOSBB110
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Includes
  • Open humidity chamber, slide rack, program card
  • Wash Module Includes
  • Four wash sleeves, slide rack, drip tray
Processes slides through incubation/thermal cycling and stringency washing of most hybridization and PCR protocols
  • Modular processing system can process up to 20 slides simultaneously
  • Slides are placed in the slide rack for easy transfer between modules
  • High-capacity, flat-block cycler with twin thermal cycling blocks can be used for simple incubations or more complex thermal cycling reactions
  • Unit minimizes intra- and inter-slide temperature variability to ensure assay reproducibility
  • Integral, removable humidity chamber preserves sample integrity
  • Two temperature control modes provide accurate temperature control
    • Simulated Slide Control constantly calculates the sample temperature and adjusts the block temperature as necessary
    • Block Control monitors the actual block temperature
Practice of the patented Polymerase Chain Reaction (PCR) process requires a license in certain countries. The Thermo Scientific Thermal Cycler is an Authorized Thermal Cycler and may be used with PCR licenses available from Applied Biosystems. Its use with Authorized Reagents also provides a limited PCR license in accordance with the label rights accompanying such reagents.