Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersil GOLD™ aQ Javelin™ Guard HPLC Columns

Catalog No. 03051246 Shop All Thermo Scientific Products
Change view
Click to view available options
Particle Size:
5 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size
03-051-246 5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-051-246 Supplier Thermo Scientific™ Supplier No. 25305012106
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns

  • Direct-connection, patented fingertight design has a 1/16 in. CPI tip that attaches directly to the inlet of the HPLC column with tubing or wrenches
  • Requires no holder, allowing quick and easy replacement

Hypersil GOLD aQ columns provide enhanced retention and resolution of polar analytes.

  • Polar endcapped C18 phase for alternative selectivity
  • Retention and resolution of polar analytes
  • Ideal for highly aqueous mobile phase
  • Excellent peak shape

Order Info

Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase aQ
Product Line Hypersil GOLD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.