Learn More
Thermo Scientific™ IntelliStack™ Incubator Shaker Accessories
Catalog No. 01257882
Change view
Click to view available options
Product Type:
Humidity and Drain Carboy Kit
Stand
Window Cover
For Use With (Equipment):
IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers
IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers w/ Drawers and Closed Sides
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
01-257-882 Stand IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers
01-257-881 Humidity and Drain Carboy Kit IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers
01-257-880 Window Cover IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers
01-257-879 Stand IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers w/ Drawers and Closed Sides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 01-257-882 Supplier Thermo Scientific™ Supplier No. 88900000
Add to Cart
Edge
Add to Cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
The Thermo Scientific™ IntelliStack™ accessories are designed to work with the IntelliStack Incubator Shaker and IntelliStack CO2 Incubator Shaker.
  • IntelliStack™ Incubator Accessories are compatible with both the CO2 Incubator Shaker and the Non-CO2 Incubator Shaker.
  • The IntelliStack Stands are available with either an open support design (88900000) or with two drawers for added storage (88900003). Both stand options are 34 cm in height and can be used with one or two Shakers.
  • The IntelliStack Window Cover is a removable metal visor that attaches to the shaker to protect the interior for ambient light (88900002).
  • The IntelliStack Humidity and Drain Carboy Kit maximize the humidity control capabilities of the IntelliStack Shaker.
TRUSTED_SUSTAINABILITY
Specifications
Holds Supports up to 2 IntelliStack Shakers
Product Type Stand
Dimensions (L x W x H) 34.9 x 55.1 x 26.8 in. (886 x 1400 x 680 mm)
For Use With (Equipment) IntelliStack CO2 Incubator Shakers and IntelliStack Incubator Shakers