Learn More
Thermo Scientific™ Low Profile Tubes and Domed Caps, strips of 8, assorted colors
Shop All Thermo Scientific ProductsDescription
The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 μL.
The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.
Highlights
- Suitable for 0.2 mL thermal cycler blocks
- 150 μL maximum tube capacity when capped
- Low profile to reduce dead space and increase PCR efficiency
- Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
- PCR
- qPCR/Real-Time PCR
Hazardous: No
Storage Condition: Ambient
PCR, qPCR and Real-Time PCR.
Certifications
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
| Certifications/Compliance | Certified DNA/RNase Free |
| Color | Assorted |
| Volume (Metric) Working | 0.1 mL |
| Volume (Metric) | 0.15 mL |
| Quantity | 250 strips |
| For Use With (Equipment) | Thermal Cyclers |
| Material | Polypropylene |
| Reaction Speed | Low profile |
| Closure Type | Domed Cap |
| Closure Included | Dome Caps |
| Show More |
Frequently Asked Questions (FAQs)
We recommend Thermo Scientific low profile PCR plates and tubes for reaction volumes below 20 µL. The low profile design minimizes the airspace between the PCR reaction and reduces evaporation effects, thereby increasing PCR efficiency in low volume reactions.
Low profile Thermo Scientific PCR plastics have shorter wells than the standard profile plastics, decreasing the “dead space” between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.
They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.
Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.
Please select your real-time PCR (qPCR) or thermal cycler (PCR) instrument model from the drop-down menu in the qPCR and PCR Plastics Selection Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html) to see the list of all validated plastic consumables (plates, tube strips, single tubes, caps, seals, adhesive films, and accessories) we offer that are compatible with your instrument.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.