Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ HDPE, Carboy

Catalog No. 12566112
Change view
Click to view available options
2.6L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
12-566-112 2.6L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-566-112 Supplier Thermo Scientific™ Supplier No. 4714

For Use With: For collection, transport, and storage of lab specimens; Color: White

  • 2.6L
  • Nonsterile

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 0.68 gal.
Capacity (Metric) 2.6 L
Color White
For Use With (Application) For collection, transport, and storage of lab specimens
Material HDPE
Product Type Carboy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.