Learn More
Thermo Scientific™ Nunc™ Open Well Upper Structures
Catalog No. 12565600 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 100
Catalog No. Quantity
12-565-600 Case of 100
1 options
This item is not returnable. View return policy
Catalog No. 12-565-600 Supplier Thermo Scientific™ Supplier No. 256665
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Versatile units in a choice of two materials
Used for bonding to a specialty base material, holding PCR plates, adding agar to each well, or other purposes
  • Nonsterile, black
  • 96- well formats available
  • Polystyrene
  • Nonsterile
  • Without lid
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
No. per Case 100
Plate Compatibility 96-Well Plate
Quantity Case of 100