Learn More
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific™

Description
Eliminate spills and ensure easy cleanup using calibration buffers in the pouch.
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
- Manufactured under ISO 9000 quality standards
- Pouches are 15mL each and are available in 10 packs or 25 packs
- Buffers are also available in pint (475mL) bottles, portable five packs of 2 oz. (5 x 60mL) bottles, convenient All-in-One buffer kits and 5 gallon (19L) cubitainers
Specifications
Specifications
| pH | 4.01 |
| Quantity | 10 Pouches |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.