Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orion™ pH Buffer Bottles, Thermo Scientific™
SCI_ED

Catalog No. s04973
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04973 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S04973 Supplier Thermo Scientific Supplier No. 916099/EMD
This item is not returnable. View return policy

Perform accurate, reproducible pH calibrations

All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability. Manufactured under ISO 9000 quality standards.

Buffer Color Codes:

  • pH 4.01 buffer is pink
  • pH 7.00 buffer is yellow
  • pH 10.01 buffer is blue

The buffers are also available in pint (475mL) bottles, portable five packs of 2 oz. (5 x 60mL) bottles, 5 gallon (19L) cubitainers and single-use pouches.

TRUSTED_SUSTAINABILITY

Specifications

Description All-in-One Buffer Kit
For Use With (Application) Accurate, reproducible pH calibrations
pH 4.01, 7.00, 10.01
Quantity 60 mL
Traceability to NIST Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.