Learn More
Thermo Scientific™ Owl™ ADJ2 Adjustable System Accessories
Catalog No. OWADJ2GK Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Gasket
Catalog No. Type
OW-ADJ2GK Gasket
1 options
Catalog No. OW-ADJ2GK Supplier Thermo Scientific™ Supplier No. ADJ2GK
Designed for increased resolving distance on a limited number of samples
  • Extremely versatile adjustable system.
  • The height of the upper buffer chamber (UBC) may be adjusted from 20 to 70cm by sliding the UBC up or down the two support poles.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ADJ1 Giraffe™ Adjustable Gel Electrophoresis System
Type Gasket