Learn More
Thermo Scientific™ Owl OSP Electrophoresis Power Supplies
Catalog No. OWOSP500 Shop All Thermo Scientific Products
Change view
Click to view available options
Voltage:
10-300 V, in 1 V steps
10-500 V, in 1 V steps
Model:
OSP-300
OSP-300 Lightning Volt
OSP-500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Model
OW-OSP-500 10-500 V, in 1 V steps OSP-500
OW-OSP300HV 10-300 V, in 1 V steps OSP-300
OW-OSP-300 10-300 V, in 1 V steps OSP-300 Lightning Volt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. OW-OSP-500 Supplier Thermo Scientific™ Supplier No. OSP500
Compact, easy-to-use 10-300V and 10-500V power supply units
  • Built-in timer permits unattended runs
  • Four outputs allow four units to be run with clear LED readout
  • Safety features include no-load detection, current/load alarm, and sudden load detection
  • Samples are protected by automatic restart after power failure
  • 300V unit ideal for DNA, RNA, mini SDS-page, pulse field electrophoresis, tank electroblotting, and electroelution
  • 500V unit suited for DNA, RNA, and protein electrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Model OSP-500
Voltage 10-500 V, in 1 V steps