Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Owl™ P8DS Dual Gel Electrophoresis System
SCI_ED

Catalog No. s98618 Shop All Fisherbrand Products
Change view
Click to view available options
P8DS dual gel electrophoresis system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S98618 P8DS dual gel electrophoresis system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98618

Fast run times and easy operation for excellent results.

Manufactured using acrylic fabrication technology, providing a durable, long-lasting system. The design enables a very simple clamping mechanism that makes loading glass cassettes and most manufacturers' precast gels trouble-free.

  • Quickly and easily cast and run one or two gels in the same device
  • Unique agarose plug method makes casting gel simple and quick
  • Gel assemblies do not have to be moved once set up
  • Ideal for second dimension protocols of 2-D electrophoresis, SDS and agarose gels and native PAGE electrophoresis
  • 8 to 10L × 10cmW gel size

  • 13L × 17W × 14cmH footprint
  • 150mL upper buffer chamber running buffer
  • 150mL lower buffer chamber running buffer
  • 300mL total running buffer

TRUSTED_SUSTAINABILITY

Specifications

Includes Buffer chamber with internal cooling core, lower Buffer chamber and SuperSafe™ lid (with attached power supply Leads)
Volume (English) Buffer 10.14 oz.
Volume (Metric) Buffer 300 mL
Warranty 3 Years
Dimensions (L x W x H) 6.7 x 5.1 x 5.5 in. (17 x 12.95 x 14 cm)
Length (English) 6.7 in.
Length (English) Gel 3.14 to 3.93 in.
Length (Metric) 17 cm
Length (Metric) Gel 8 to 10 cm
Width (English) 5.1 in.
Width (English) Gel 3.93 in.
Width (Metric) 12.95 cm
Width (Metric) Gel 10 cm
Height (English) 5.5 in.
Height (Metric) 14 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.