Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Owl™ Separation HEP-1 Semi Dry Electroblotting System
SCI_ED

Catalog No. S98622 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Electroblotting System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S98622 Electroblotting System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98622

Includes

Includes: base with stainless steel cathode plate, lid with platinum/titanium anode plate, power supply leads, heavy duty knobs

Sleek, low profile systems for fast, efficient transfer of proteins and nucleic acids to membranes.

System requires only minimal amounts of buffer (about 250mL). The buffer contained within the blotting filter paper is sufficient for efficient transfer of molecules.

  • Base with stainless steel cathode plate
  • Lid contains a platinum/titanium anode plate
  • 20L x 20cmW transfer area allows up to 4 mini gels
  • 45L x 35cmW transfer area allows up to 8 mini gels or a variety of larger size gels
  • Transfers may be done in as little as 15 minutes
  • Ideal for proteins less than 150kd
  • 25L x 25W x 5cmH footprint

TRUSTED_SUSTAINABILITY

Specifications

Device Holds 4 MiniGels or other size Gels totaling 20 x 20cm
Type Electroblotting System
Includes Base, lid, power supply Leads, heavy duty knob
Dimensions (L x W x H) 9.8 x 9.8 x 2 in. (25 x 25 x 5 cm)
Length (English) 9.8 in.
Length (English) Gel 7.8 in.
Length (Metric) 25 cm
Length (Metric) Gel 20 cm
Width (English) 9.8 in.
Width (English) Gel 7.8 in.
Width (Metric) 25 cm
Width (Metric) Gel 20 cm
Height (English) 2 in.
Height (Metric) 5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.