Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion U Multiplex PCR Master Mixes

Catalog No. p-4913485
Change view
Click to view available options
Color:
Colorless
Green
No. of Reactions:
100 Reactions
500 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Reactions
F564S Green 100 Reactions
F562L Colorless 500 Reactions
F562S Colorless 100 Reactions
F564L Green 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. F564S Supplier Thermo Scientific™ Supplier No. F564S
Only null left
Add to Cart
Add to Cart

Thermo Scientific Phusion U Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube.

Thermo Scientific Phusion U Green Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube. Over 20 primer pairs may be combined into a single reaction for highly specific and efficient multiplexing over a broad range of primer and template concentrations.

The Phusion U Multiplex PCR Master Mix contains a proprietary buffer with balanced concentrations of all PCR components eliminating the need for tedious optimization. The Green format further simplifies the workflow - it includes a density reagent and two electrophoresis tracking dyes for direct loading of PCR products on gels.

Highlights

  • Specific and efficient multiplex PCR of over 20 targets up to 2.5 kb
  • Excellent results with templates of suboptimal purity due to inhibitor tolerance
  • Fast cycling and direct loading of PCR products on gels

Applications

  • Genotyping
  • Pathogen detection
  • Food testing
  • Analysis of genetically modified organisms
  • Amplification of microsatellites

The master mix is based on Phusion U Hot Start DNA Polymerase, a high performance enzyme developed using fusion technology. Similarly to other enzymes from Phusion family, Phusion U DNA Polymerase incorporates more nucleotides per binding event than any other DNA polymerase. This allows achieving high yields of PCR products with shorter extention times and consequently reduced total protocol times. Significantly enhanced processivity also enables successful multiplex PCR on difficult targets such as GC-rich templates or templates of suboptimal purity. Being highly tolerant of many PCR inhibitors, Phusion U Multiplex PCR Master Mixes can even be used with unpurified samples such as blood or serum.

Specificity of multiplex PCR is increased by Affibody-mediated hot start mechanism. Reversibly bound Affibody ligand inhibits the polymerase at ambient temperatures preventing amplification of nonspecific products and formation of primer-dimers.

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • Phusion U Hot Start DNA Polymerase
• 2X reaction buffer
• 3 mM MgCl2
• 400 μM of each dATP, dTTP, dCTP and dGTP

Master mix is supplied with nuclease-free water.
Store at -20°C.
GC-Rich PCR Performance High
Polymerase Phusion U Hot Start DNA Polymerase
Reaction Speed Fast
Product Type Multiplex PCR Master Mix
Quantity 100 x 50 μL Reactions
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 25X
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format SuperMix or Master Mix
Size (Final Product) 2.5 kb or less
Color Green
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.