Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TMT10plex™ Isobaric Label Reagents and Kits
GSA_VA

Catalog No. PI90111
Change view
Click to view available options
Quantity:
1 x 0.8 mg (per tag), 10 Reactions
1 x 5 mg (per tag), 60 Reactions
3 x 0.8 mg
3 x 0.8 mg (per tag), 30 Reactions
8 x 0.2 mg
8 x 40 μg​ (per tag), 80 Reactions
Product Type:
Isobaric Label Reagent Set
Isobaric Mass Tag Labeling Kit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PI90111 3 x 0.8 mg (per tag), 30 Reactions Isobaric Label Reagent Set
PI90110 1 x 0.8 mg (per tag), 10 Reactions Isobaric Label Reagent Set
PI90406 1 x 5 mg (per tag), 60 Reactions Isobaric Label Reagent Set
PI90309 8 x 0.2 mg Isobaric Label Reagent Set
PI90113 3 x 0.8 mg Isobaric Mass Tag Labeling Kit
FERA58332 8 x 40 μg​ (per tag), 80 Reactions Isobaric Label Reagent Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. PI90111 Supplier Thermo Scientific™ Supplier No. 90111
Add to Cart
Add to Cart

TMT10plex Isobaric Label Reagents and Kits contain ten isobaric compounds with the same mass and chemical structure (i.e., isotopomeric) composed of an amine-reactive NHS-ester group, a spacer arm, and a mass reporter.

TMT10plex Isobaric Label Reagents and Kits contain ten isobaric compounds with the same mass and chemical structure (i.e., isotopomeric) composed of an amine-reactive NHS-ester group, a spacer arm, and a mass reporter. Up to ten different peptide samples prepared from cells or tissues can be labeled in parallel and then combined for analysis. For each sample, a unique reporter mass (i.e., TMT10 126-131 Da) in the low-mass region of the high-resolution MS/MS spectrum is used to measure relative protein expression levels during peptide fragmentation and tandem mass spectrometry.

The Tandem Mass Tag (TMT) Isobaric Label reagents and kits are specifically designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry. The TMT10plex label reagents share an identical structure with TMTzero, TMTduplex, and TMTsixplex reagents, but contain different numbers and combinations of 13C and 15N isotopes in the mass reporter. The different isotopes result in a 10-plex set of tags that have mass differences in the reporter that can be detected using high resolution Orbitrap MS instruments.

Advantages of the TMT10plex label reagents include increased multiplex relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents are ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or improving statistics on biological replicates.

Features of TMT10plex Isobaric Label reagent sets and mass tagging kits:

  • Powerful—multiplexed MS analysis increases sample throughput and enables relative quantitation of up to ten different samples derived from cells, tissues, or biological fluids
  • Consistent—identical reagent structure and performance among TMTzero, TMTduplex, TMTsixplex, and TMT10plex reagents allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research across platforms and datasets
  • Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
  • Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
  • Compatible—optimized for use with high resolution Orbitrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3.

Applications:

  • Protein identification and quantitation from multiple samples of cells, tissue, or biological fluids
  • Protein expression profiling of normal vs. disease states or control vs. treated
  • Quantitative analysis of proteins for which no antibodies are available
  • Identification and quantitation of membrane and post-translationally modified proteins
  • Identification and quantification of hundreds to thousands of proteins in a single experiment

TMT Reagents are provided as standalone sets, a 96-well format, or in optimized kit formats containing all necessary reagents and controls for maximum flexibility, convenience, and reliability. When combined with the industry-leading, high resolution Thermo Scientific Orbitrap instruments and software, TMT10plex reagents provide integrated total solutions for quantitative protein expression analysis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Isobaric Label Reagent Set
Product Line TMT10plex
Sufficient For 30 Reactions
Content And Storage Sufficient for: Three 10-plex (3 x 10-way) experiments
• TMT10-126 Label Reagent, 3 x 0.8 mg
• TMT10-127N Label Reagent, 3 x 0.8 mg
• TMT10-127C Label Reagent, 3 x 0.8 mg
• TMT10-128N Label Reagent, 3 x 0.8 mg
• TMT10-128C Label Reagent, 3 x 0.8 mg
• TMT10-129N Label Reagent, 3 x 0.8 mg
• TMT10-129C Label Reagent, 3 x 0.8 mg
• TMT10-130N Label Reagent, 3 x 0.8 mg
• TMT10-130C Label Reagent, 3 x 0.8 mg
• TMT10-131 Label Reagent, 3 x 0.8 mg

Store at –20°C.

Label or Dye Tandem Mass Tag™ (TMT) Labels
Starting Material Cell Lysate
Final Product Type Protein
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 3 x 0.8 mg (per tag), 30 Reactions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.