Learn More
Thermo Scientific™ Pierce™ IQ Assay Kits
Catalog No. PI62520
Change view
Click to view available options
:
Calibrator Set For Iq Kinase
Catalog No.
PI62520 Calibrator Set For Iq Kinase
1 missing translation for 'options'
Catalog No. PI62520 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 62520
Includes
Includes:
  • Dye-labeled substrate
  • IQ Reagent A
  • IQ Reagent B
A direct, noncompetitive assay system for kinase or phosphatase activity
Quantify kinase or phosphatase activity by direct measurement of the phosphorylation state of a fluorophore-labeled peptide substrate. Phosphorylation is measured by the change in fluorescence intensity that occurs when a proprietary iron-containing compound binds specifically to phosphoryl groups on the peptides. The change in fluorescence is proportional to the extent of phosphorylation of the fluorophore-labeled peptide, because the iron-containing compound quenches the fluorescent emission exclusively on the phosphorylated form of the fluorophore-labeled peptide.
  • Homogeneous, in-solution assay format does not require wash steps or beads
  • Universal system for any kinase or phosphatase—no antibody required
  • Assay is directly scalable from a 96-well format to 384- and 1536-well formats
  • Robust results—Z« of 0.6 for 10% conversion of substrate in a kinase reaction and Z« of 0.7 for 10% conversion of substrate in a phosphatase reaction
  • Addition of IQ reagents stops the enzyme reaction, so plates may be read immediately or up to two days later
  • Results can be read on any fluorometer
†Indicates number of assays performed in a 96-well format.