Learn More
Thermo Scientific™ Quick Connect Luer Adapters
Description
Adapters conform to ISO requirements for medical luer taper configuration and performance (45 psi/3.1 bar).
Specifications
Specifications
| Type | Luer Adapter |
| Material | ETFE |
| Diameter (Metric) Inner | 1.3 mm |
| For Use With (Equipment) | Female Luer to 1/4-28 Male |
| Length (Metric) | 20.8 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.