Learn More
Thermo Scientific™ RadEye PRD Personal Radiation Detector
Catalog No. 19155225
Change view
Click to view available options
Description:
RadEye PRD Safety Kit, detector calibrated in R/h, with 59in. extension
RadEye PRD Safety Kit, detector calibrated in Sv/h, with 157in. extension
RadEye PRD Safety Kit, detector calibrated in Sv/h, with 59in. extension
RadEye PRD Safety Kit,detector calibrated in R/h, with 157in. extension
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
19-155-225 RadEye PRD Safety Kit,detector calibrated in R/h, with 157in. extension
19-155-224 RadEye PRD Safety Kit, detector calibrated in R/h, with 59in. extension
19-155-226 RadEye PRD Safety Kit, detector calibrated in Sv/h, with 59in. extension
19-155-227 RadEye PRD Safety Kit, detector calibrated in Sv/h, with 157in. extension
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-155-225 Supplier Thermo Scientific™ Supplier No. 425067190157
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Removable rubber sleeve for extra protection.
High sensitivity with high selectivity
Lightweight, handheld radiation detector can be used as a search and survey tool. The instrument can be used in a belt-loop holster for hands-free operation.
  • Menu-driven with an intuitive, easy-to-use format
  • 5000 to 10,000 times more sensitive than a typical electronic dosimeter; incorporates highly sensitive NaI(TI) scintillation detector with miniature photomultiplier for detection of very low radiation levels (emphasis on gamma emissions below 400 KeV)
  • Patented Natural Background Rejection (NBR) technology provides high sensitivity with high selectivity; eliminates nuisance alarming due to naturally recurring radiation sources (such as granite)
  • Low-power-technology components and fully automated self-checks result in minimum maintenance
  • Equipped with large, clear graphic display and audible/visible/vibrating alarms; earphone-output for silent alarming
RadEye PRD Safety Kits include a PDR Personal Radiation Detector plus:
  • Holster for RadEye PRD
  • Universal RadEye PRD “snap in” adapter
  • RadEye PRD Test Adapter (natural lutetium oxide)
  • Short handle for “snap in” of the universal RadEye adapter
  • Extension (59 or 157 in.)
  • USB to the RadEye PRD's IR port adapter cable
  • RadEye desktop stand with sounting support of the adapter cable
  • Special application-specific software and handbook
Compliance
Designed to meet ANSI™ 42.33/1, 42.32 and IEC 62401.
TRUSTED_SUSTAINABILITY
Specifications
Description RadEye PRD Safety Kit,detector calibrated in R/h, with 157in. extension