Learn More
Thermo Scientific™ Remel™ Demi-Fraser Broth Base
Catalog No. R453466 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 kg
Catalog No. Quantity
R453466 10 kg
1 options
This item is not returnable. View return policy
Catalog No. R453466 Supplier Thermo Scientific™ Supplier No. R453466
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
For cultivation of Listeria monocytogenes.
Requires addition of Fraser Broth Additive, Part No. 450361.
Order Info
Requires addition of Fraser Broth Additive, Part No. R450361.
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Media Type Demi-Fraser Broth Base
Quantity 10 kg
For Laboratory Use Only