Learn More
Thermo Scientific™ Remel™ GN Broth
Catalog No. R08393
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R08393 20/Pk.
R08383 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R08393 Supplier Thermo Scientific™ Supplier No. R08393
A liquid medium recommened for use in qualitative procedures for selective enrichment of enteric gram-negative bacilli
TRUSTED_SUSTAINABILITY
Specifications
Fill Volume 8 mL
Quantity 20/Pk.