Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Repipet™ II Dispensers

Catalog No. 1368762B Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-687-62B 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-687-62B Supplier Thermo Scientific™ Supplier No. 9010

missing translation for 'validMainframeMsg'

Tough fluorocarbon resin sleeve entirely protects dispensing mechanism

Suitable for use with all lab reagents except hydrofluoric acid and organic solvents.

  • Dispensing mechanism features extra-thick borosilicate glass barrel and plunger
  • Included integrated 100-division scale
  • PTFE inlet and outlet tips
  • With amber glass bottles; 500mL for 5mL dispenser and 1250mL bottle for 10 and 20mL dispensers
  • Universal models are available for use with user-supplied bottle (amber bottle not included); include three caps/liners (28-430, 33-430, and 38-430) and 12 in. (30.5cm) of PTFE resin tubing
  • Magnifying indicator allows easy and accurate viewing of scale
  • Reproducibility: ±0.2%
  • Volume range: 0.01 to 20mL
  • Accuracy: ±1.5%

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.