Learn More
Epredia™ Cassette 1 Biopsy Cassettes
Catalog No. 1000969 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Pink
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
10-009-69 Green
10-010-90 Pink
10-010-91 Blue
10-009-68 White
10-009-70 Yellow
10-009-71 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10-009-69 Supplier Epredia™ Supplier No. 1000969
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specially designed to hold biopsy specimens during processing and embedding
  • Made of acetal polymer
  • Cassette writing face is at a 30° angle
  • Pore style is small square
  • Locking lid attached by two hinges
  • Supplied in a box of 500 cassettes with attached breakaway lids
Order Info
Not available in Tube Packs
Specifications
Pore Style Square
Writing Face Angle 30°
Closure Type Hinged
Color Green
Material Acetal Polymer
Product Type Biopsy Cassette