Learn More
Epredia™ Shandon™ Electronic Floor Scale
Catalog No. 20560 Shop All Epredia Products
Change view
Click to view available options
Type:
Digital Indicator
Floor Scale
Quick Pit Frame
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
20560 Floor Scale
20565 Quick Pit Frame
20570 Digital Indicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20560 Supplier Epredia™ Supplier No. 20560/DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Crafted from epoxy-painted mild steel
  • A complete line of of epoxy-painted mild steel, electronic floor scales are available
  • Variety of sizes are available from 3 × 3 ft. (91.4 × 91.4cm) to 5 × 7 ft. (152.4 × 213.4cm)
  • Ramped models are also available
  • Variety of capacities are available from 1000 to 10,000 lb. (453.6 to 4536kg)
  • No mechanical adjustment is required
  • Hermetically sealed load-sensing elements allow the user to clean the unit without damaging the weighing mechanisms
  • Standard scale is installed into a pit, but ramped and freestanding units are available
  • Pimarily constructed of stainless steel, including the load cells, weighbridge, scale frame, optional ramps
Order Info
Scale 20560 does not include pit frame, ramp, or digital indicator (sold separately)
Certifications
A Certificate of Conformance, Number 91-097 was issued under the National Type Evaluation Program (NTEP).
Compliance
Scales meet /exceed the Class III, 5000 division accuracy with the National Institute of Standards and Technology (NIST™) Handbook 44.
Specifications
Capacity 453.6 to 4536 kg
Type Floor Scale