Learn More
Thermo Scientific™ Stand-alone Guard Cartridge Holders
Catalog No. 84300 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
4 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
84300 3 mm
84000 4 mm
84200 2.1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 84300 Supplier Thermo Scientific™ Supplier No. 84300
Eliminate the requirement for extra fittings with Thermo Scientific Guard Cartridge Holders.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 3 mm