Learn More
Thermo Scientific™ SUN-SRi™ Calibrated To Deliver Non-Standard Size Volumetric Pipets
Description
All pipets have standard size tip openings.
Specifications
Specifications
| Calibration | To Deliver |
| Certifications/Compliance | ASTM E969 |
| Class | A |
| Material | Borosilicate Glass |
| Volume (Metric) | 1.25 mL |
| Type | Volumetric Pipet |