Learn More
Thermo Scientific™ Graphite/Vespel Ferrule for Transfer Line
Catalog No. 290SG020 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk
Catalog No. Quantity
29-0SG-020 10/Pk
1 options
Catalog No. 29-0SG-020 Supplier Thermo Scientific™ Supplier No. 290SG020
Add to Cart
Add to Cart
Easy to install for quick column changeovers
The Thermo Scientific SWAP-IT GCMS Interface provides a powerful tool for switching capillary columns without the need to vent the MS.
  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Description Graphite/Vespel Ferrule; For Thermo Scientific Transfer Line
Quantity 10/Pk
For Use With (Equipment) DSQ, DSQ II, PolarisQ, ITQ