Learn More
Thermo Scientific™ TRACE™ TR-35MS GC Columns
Catalog No. 03151625 Shop All Thermo Scientific Products
Catalog No. 03-151-625 Supplier Thermo Scientific™ Supplier No. 260C131P
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Mid-polarity columns excellent for many applications
  • Mid-polarity phase, 35% Phenyl polysilphenylene-siloxane
  • Exceptionally low surface activity
  • Low bleed even at elevated temperatures
  • Similar to DB-35, DB-35MS, HP-35, HP-35MS,MDN-35, Rtx-35, SPB-35, BPX35
Pesticides, Herbicides, Drugs of Abuse, Poly Aromatic Hydrocarbons, Pharmaceuticals