Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Versette ClipTips™

Catalog No. 14387574
Change view
Click to view available options
Model:
12.5μL
125/300μL
1250μL
30μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Tip Style:
Automation
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Filtered Sterility Tip Style
14-387-574 125/300μL Filtered Sterile Automation
14-387-571 30μL Filtered Sterile Automation
14-387-570 12.5μL Filtered Sterile Automation
14-387-572 30μL Non-filtered Sterile Automation
14-387-573 30μL Non-filtered Non-sterile Automation
14-387-579 1250μL Non-filtered Non-sterile Automation
14-387-575 125/300μL Non-filtered Sterile Automation
14-387-578 1250μL Non-filtered Sterile Automation
14-387-577 1250μL Filtered Non-sterile Automation
14-387-576 125/300μL Non-filtered Non-sterile Automation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14-387-574 Supplier Thermo Scientific™ Supplier No. 5708

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Unique interface locks tip onto Versette automated liquid handling system.

Tips have a clasp or clip that locks in place to ensure an effective seal and will only release when mechanically ejected.

  • Unique design – On or Off only – tips lock in place providing a secure fit and preventing loose, uneven or leaky tips
  • Minimizing cross contamination between samples, the tips reach the bottom of tubes
  • Versatile tip range with three sizes for volumes: 1250μL, 300/125μL and 30/12.5μL
  • Rigid rack allows stable, even tip seating

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 to 125 μL
Filtered Filtered
Sterility Sterile
Model 125/300μL
For Use With (Application) On or off tips lock in place providing a secure fit and preventing loose, uneven or leaky tips, minimizing cross contamination between samples, provide stable, straight mounting every time
Format Rack
Tip Style Automation
For Use With (Equipment) Thermo Scientific Versette
Product Line Versette ClipTips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.