Learn More
Thermo Scientific™ Versette ClipTips™
Description
Tips have a clasp or clip that locks in place to ensure an effective seal and will only release when mechanically ejected.
- Unique design – On or Off only – tips lock in place providing a secure fit and preventing loose, uneven or leaky tips
- Minimizing cross contamination between samples, the tips reach the bottom of tubes
- Versatile tip range with three sizes for volumes: 1250μL, 300/125μL and 30/12.5μL
- Rigid rack allows stable, even tip seating
Specifications
Specifications
| Volume (Metric) | 10 to 125 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Model | 125/300μL |
| For Use With (Application) | On or off tips lock in place providing a secure fit and preventing loose, uneven or leaky tips, minimizing cross contamination between samples, provide stable, straight mounting every time |
| Format | Rack |
| Tip Style | Automation |
| For Use With (Equipment) | Thermo Scientific Versette |
| Product Line | Versette ClipTips |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.