Learn More
Thermo Scientific™ ViewPoint Enterprise Plug-Ins and Software
Catalog No. 19175555
Change view
Click to view available options
Product Type:
Plume Tracker Simulator Viewpoint Plug-In
Viewpoint Data Management Server
Viewpoint Enhanced Survey Software
Viewpoint Enterprise Input Device Plug-Ins
Viewpoint Enterprise Plug-In
Viewpoint Enterprise Plug-Ins
Viewpoint Enterprise Software
Viewpoint Server Software
Description:
1 lot
Customer supplied server plugins not included
Logging Server and reporting tool
Per 10 unit Package
Per 100 unit Package
Per 50 unit Package
Per unit
Third-party; Per 100 unit Package
Third-party; Per Package
Third-party; Sold each
To interface with all probes of FH 40
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Description
19-175-555 Viewpoint Server Software Customer supplied server plugins not included
19-175-210 Viewpoint Enterprise Plug-Ins Third-party; Sold each
19-175-217 Viewpoint Enterprise Input Device Plug-Ins Per 50 unit Package
19-175-218 Viewpoint Enterprise Input Device Plug-Ins Per unit
19-175-219 Viewpoint Enterprise Input Device Plug-Ins Per 100 unit Package
19-175-213 Viewpoint Enterprise Input Device Plug-Ins Per 10 unit Package
19-175-212 Viewpoint Enterprise Plug-Ins Third-party; Per Package
19-175-215 Viewpoint Enterprise Plug-Ins Third-party; Per 100 unit Package
19-175-211 Viewpoint Enterprise Plug-Ins Third-party; Per Package
19-175-214 Viewpoint Enterprise Input Device Plug-Ins Per unit
19-175-216 Viewpoint Enterprise Input Device Plug-Ins Per unit
19-175-227 Viewpoint Enterprise Plug-In Per unit
19-175-226 Viewpoint Enterprise Plug-In Per 10 unit Package
19-177-224 Viewpoint Enterprise Software 1 lot
19-177-225 Viewpoint Enterprise Software 1 lot
19-175-550 Viewpoint Enterprise Plug-In To interface with all probes of FH 40
19-175-556 Viewpoint Data Management Server Logging Server and reporting tool
19-175-557 Viewpoint Enhanced Survey Software -
19-175-558 Plume Tracker Simulator Viewpoint Plug-In -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 19-175-555 Supplier Thermo Scientific™ Supplier No. SYS/1/70850/001
Requires
System Specifications
Operates with Windows™ NT/2000/XP
Client Software Requirements
  • Microsoft Windows™ NT 4 Service Pack 6 or
  • Microsoft Windows™ 2000 Service Pack 4 or
  • Microsoft Windows™ XP Service Pack 1 or higher
Plug-ins and Software to be used with the ViewPoint Appliance. The platform is the base for a complete customizable solution for remote monitoring and remote real-time command and control; capable of integrating radiation, environmental, and general purpose detectors.
Plug-Ins
Each instrument, detector or sensor, which is capable of being remotely monitored by ViewPoint, has a unique Input Device Plug-In. This capability allows the instrument to communicate with ViewPoint Data Engine/System.
Client Plug-Ins
The ViewPoint User Interface and Data Engine are compatible with third-party Remote Monitoring Software systems. Compatability includes such systems as: MGPI-Teleview™, Industrial Innovative Solutions (IIS) – RADS™, and Bartlett – RMS™, and ESRI ArcView™ version 3.3 and 8. All purchased plug-ins are preloaded on the ViewPoint Appliance prior to delivery. Additional Input Device plug-ins, Client plug-ins, and User/Workstation License Bundles can be purchased and added to the original ViewPoint appliance via a patch file and updated security key
  • Real-time personnel dosimetry and data management
  • Area monitoring
  • Integrated remote monitoring
  • Instrument management
  • First Responder environmental monitoring
  • Site Process Monitoring
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viewpoint Server Software
Description Customer supplied server plugins not included
Operating System Windows™ NT/2000/XP
For Use With (Application) Real time personnel dosimetry and data management, area monitoring, integrated remote monitoring, instrument management, first responder environmental monitoring, site process monitoring