Learn More
Thermo Scientific™ Wide-Mouth Septa Jars; Type III glass bottle, white PP open top closure with PTFE/silicone septum
Description
- Type III soda-lime glass bottle, clear or amber
- Amber jars are recommended for light-sensitive analytes
- White polypropylene cap with bonded PTFE-faced silicone septa
- Caps assembled onto bottles
- Wide-mouth design is ideal for soil samples and viscous liquids
Includes:
Certified jars come with the following features:
- Certified to meet or exceed EPA performance-based standards for volatile organic analysis
- Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
- Each bottle individually barcoded with lot number and unique container number
- Analysis labels included unattached for optional use
- Carton sealed with security tape providing tamper evidence
- Chipboard divider helps protect jars from breakage; requires additional over-pack for shipping
Unprocessed jars are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; requires additional over-pack for shipping.
Compliance
Certified and Processed jars meet or exceed EPA performance based standards for volatile organic analysis; manufactured under an ISO certified quality management system.
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Color | Clear |
| Material | Soda Lime Glass |
| Product Type | Septum Processed |
| Closure Size | 48-400 mm |