Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Thermoelectric Demonstrator
SDPSCI_ED

Catalog No. S43684 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
9 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43684 9 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43684 Supplier United Scientific Supplies Supplier No. TED001
Add to Cart
Add to Cart

Demonstrates the Second Law of Thermodynamics.

  • Immerse one of the aluminum legs into ice cold water and the other leg into hot water to demonstrate how heat transfer can be converted to electrical energy.
  • This conversion turns the turbine.
  • Show the reverse effect of converting electrical energy to heat energy by plugging the apparatus into a DC power supply.
  • In this activity, DC current is given to the unit so one of the strips will heat up and the other will remain cold.
  • Includes a set of two 24 in. banana connecting cords and a detailed activity guide.

Specifications

Grade 9 to 14
Format Kit
Product Type Thermoelectric Demonstrator
Languages English
Subject Physics - Thermodynamics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.