Learn More
United Scientific Supplies Thermoelectric Effects Apparatus
SDPSCI_ED
Catalog No. S07393 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Thermoelectric effects apparatus
Catalog No.
S07393 Thermoelectric effects apparatus
1 options
Catalog No. S07393 Supplier United Scientific Supplies Supplier No. THEA01
Allows the thermal behavior of a resistor, thermistor, and thermocouple to be studied, as well as a closed loop control system.
Apparatus consists of the following:
  • Oven containing a cooling fan
  • Metal block with an electric heater
  • Three samples (thermocouple, a three-pole metal resistor, and a thermistor)
  • Control system sensor
  • Control unit that provides the heating current
  • Connectors to the three sample devices
  • Digital temperature controller
  • Insulated flask to hold ice water for the cold junction of the thermocouple
The signals from the three sample devices can be read by any suitable measuring device provided by the user.
The digital control system can be operated in P, PI, PD, and PID modes to show their characteristics, and a wide range of parameters can be set
Specifications
Product Type Thermoelectric Effects Apparatus
For Use With (Application) To learn by investigating the thermal characteristics of three commonly-used electrical thermometry devices and also to learn the features and behavior of a closed loop digital temperature control system.
Includes Oven containing a cooling fan, a massive metal block with an electric heater, the three samples, and the control system sensor, and a control unit that provides the heating current, connectors to the three sample devices and the digital temperature controller. The oven and the control unit are connected by three multi-pole cables.
Weight (English) 11.2 lb.
Weight (Metric) 5.1 kg