Learn More
Thermoelectricity Demo
SDPSCI_ED
Catalog No. S96825 Shop All Science First Products
Change view
Click to view available options
Thermoelectricity demo model
Catalog No.
S96825 Thermoelectricity demo model
1 options
Catalog No. S96825 Supplier Science First Supplier No. 6154595
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Illustrate the Seebeck Effect
When a thermoelectric loop is fashioned from two dissimilar metals and they are connected at two junctions, and a temperature gradient exists, a voltage or electromotive force (EMF) is induced, which is enough to deflect a magnetic compass.
  • Aluminum strip on bottom
  • Bent copper strip on top
  • Depression underneath copper loop for placement of small magnetic compass
  • Attached handle for safety and ease of use
Specifications
For Use With (Application) Illustrate the Seebeck effect
Includes Depression underneath Copper loop, attached handle, Aluminum strip on bottom, bent Copper strip on top
Product Type Thermoelectricity Demonstration Model