Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermolabile UDG, 500 U, G-Biosciences
SCI_ED

Catalog No. S001117
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001117 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001117 Supplier G-Biosciences Supplier No. 7862264
Add to Cart
Add to Cart

Thermolabile Uracil-DNA Glycosylase (UDG), originating from the species Gadus morhua, removes uracil from DNA by hydrolyzing the N-glycosylic bond between the deoxyribose and the base leaving an AP (apurinic or apyrimidinic) site.

  • The enzyme is useful in the prevention of amplicon carry-over and is completely and irreversibly inactivated by a 10 min. incubation at 55°C.

Specifications

Packaging Vial
Enzyme Uracil-DNA Glycosylase
Sensitive to Heat Inactivation Yes
Research Category DNA Repair, Next-generation Sequencing, PCR
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.