Learn More
Sigma Aldrich Fine Chemicals Biosciences ThermolyneR Small Benchtop1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5097951EA
Encompass
Ideal for ashing organic or inorganic samples and ignition tests. Can be used for determination of volatile and suspended solids.Drop-down door convenient for loading and unloadingAnalog controlsDisplay reads chamber temperature or set-point temperatureSafety switch removes power from heating elements when door is open
Catalog No. 50-290-8589