Learn More
Thermco Products Inc Precision lab Hg thermometer, Fractional degree, -1 deg./101 deg.C, 0.1 deg.C div, 76mm imm, 610mm
Supplier:  Thermco Products Inc ACC10053
Encompass_Preferred
High precision mercury SAMA thermometer that meet or exceed the specifications of the American National Standards Institute. Specifications approve by N.I.S.T. and ASTM. Individually serialized and permanetly marked. Made with precision yellow backed tubing. SAMA No. CP45
Catalog No. NC0605432