Learn More
Cole-Parmer Digi-Sense Soil Thermometer, 36" L x 0.25" Dia; 0-200F/ -15-90C
Supplier:  Cole-Parmer 9058630
Encompass_Preferred
Our Process Grade bimetal thermometers offer a 3" and 5" dial sizeand are ideal for most rugged industrial temperature measurementapplications. The hermetically-sealed case offers protection fromweather and dust, and is guaranteed against fogging up. An inert gelis used to minimize pointer oscillation. These thermometers are guaranteed for seven years.
Catalog No. NC1552574
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.